Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NKIAMRE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572930
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572930 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572930 Supplier Invitrogen™ Supplier No. PA572930
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NKIAMRE Polyclonal Antibody for ICC/IF, IHC (F), IHC (P), ELISA

The protein encoded by this gene is a member of cyclin-dependent protein kinase (CDK) family. CDK family members are highly similar to the gene products of Saccharomyces cerevisiae cdc28, and Schizosaccharomyces pombe cdc2, and are known to be important regulators of cell cycle progression. This gene was identified as a gene absent in leukemic patients with chromosome 5q deletion. This loss may be an important determinant of dysmyelopoiesis. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NKIAMRE
Applications ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CDKL3
Gene Accession No. Q8IVW4
Gene Alias AI852479; B230379H01Rik; CDKL3; cyclin dependent kinase like 3; cyclin-dependent kinase-like 3; NKIAMRE; Nkiatre; Nkiatreb; serine/threonine kinase NKIATRE beta; serine/threonine protein kinase NKIATRE; Serine/threonine-protein kinase NKIAMRE; serine-threonine protein kinase NKIAMRE
Gene Symbols CDKL3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues 14-29 of the 50 kDa human NKIAMRE protein.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51265
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.