Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NKIAMRE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA14174
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA14174 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA14174 Supplier Invitrogen™ Supplier No. PA14174
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-4174 detects NKIAMRE in human samples. PA1-4174 has been successfully used in Western blot procedures. The PA1-4174 immunogen is a synthetic peptide corresponding to residues N(414) C N G L K E N P H C G G S V(428) of the 50 kDa human NKIAMRE. Maintain at 4ºC for 3 months or at -20ºC for longer periods. Stable for 1 year. Avoid freeze/thaw cycles.

The protein encoded by this gene is a member of cyclin-dependent protein kinase (CDK) family. CDK family members are highly similar to the gene products of Saccharomyces cerevisiae cdc28, and Schizosaccharomyces pombe cdc2, and are known to be important regulators of cell cycle progression. This gene was identified as a gene absent in leukemic patients with chromosome 5q deletion. This loss may be an important determinant of dysmyelopoiesis. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NKIAMRE
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CDKL3
Gene Accession No. Q8IVW4
Gene Alias AI852479; B230379H01Rik; CDKL3; cyclin dependent kinase like 3; cyclin-dependent kinase-like 3; NKIAMRE; Nkiatre; Nkiatreb; serine/threonine kinase NKIATRE beta; serine/threonine protein kinase NKIATRE; Serine/threonine-protein kinase NKIAMRE; serine-threonine protein kinase NKIAMRE
Gene Symbols CDKL3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues N(414) C N G L K E N P H C G G S V(428) of human NKIAMRE.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51265
Target Species Human
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.