Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKp46 (CD335), Mouse anti-Human, Clone: D2-9A5, Millipore Sigma™

Catalog No. p-7162919 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF205825 25 μg
MABF2058100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF205825 Supplier MilliporeSigma Supplier No. MABF205825UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-NKp46 (CD335), clone D2-9A5 , Cat. No. MABF2058, is a mouse monoclonal antibody that detects Natural cytotoxicity triggering receptor 1 (NKp46) and has been tested for use in and ELISA, and Flow Cytometry.

Natural cytotoxicity triggering receptor 1 (UniProt: O76036; also known as Lymphocyte antigen 94 homolog, NK cell-activating receptor, Natural killer cell p46-related protein, NK-p46, NKp46, hNKp46, CD335) is encoded by the NCR1 (also known as LY94) gene (Gene ID: 9437) in human. NK-p46 is a single-pass type I membrane protein that belongs to the natural cytotoxicity receptor (NCR) family and serves as a cytotoxicity-activating receptor that may contribute to the increased efficiency of activated natural killer (NK) cells to mediate tumor cell lysis and elimination of virus-infected cells. It is considered as a primary activating receptor mediating NK cell antiviral function. It is involved in the functional recognition of viral HA and hemagglutinin-neuraminidases. NKp46 is shown to be predominantly expressed in the nasal lavage of patients infected with respiratory viruses, but not in normal healthy subjects. Following the initiation of an NK-target cell interaction, NKp46 is reported to cluster at the cell membrane, specifically at the immune synapse where it mediates cytoskeletal rearrangement and cellular polarization. Human NK-p46 is synthesized with a signal peptide of 21 amino acids, which is subsequently cleaved off. The mature form has an extracellular domain (aa 22-258), a helical domain (aa 259-279), and a cytoplasmic domain (aa 280-304). It also has two Ig-like domains (aa 34-118 and 129-211) termed as D1 and D2. Five different isoforms of NKp46 have been described that are produced by alternative splicing. (Ref.: Shemer-Avni, Y., et al., (2017). Front. Immunol. 8 (Article 161); 1-10).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NKp46 (CD335)
Applications ELISA, Flow Cytometry
Classification Monoclonal
Clone D2-9A5
Conjugate Unconjugated
Dilution Flow Cytometry Analysis: A representative lot detected NKp46 (CD335) in Flow Cytometry applications (Shemer-Avni, Y., et. al. (2017). Front Immunol. 8:161).Flow Cytometry Analysis: 4μg/mL from a representative lot detected NKp46 (CD335) in NK92 cells transfected with human NKp46 (Courtesy of Angel Porgador, Ph.D., Faculty of Health Sciences, Ben Gurion university of the Negev, Israel).ELISA Analysis: A representative lot detected NKp46 (CD335) in ELISA applications (Shemer-Avni, Y., et. al. (2017). Front Immunol. 8:161).
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Natural cytotoxicity triggering receptor 1;Lymphocyte antigen 94 homolog;NK cell-activating receptor;Natural killer cell p46-related protein;NK-p46;hNKp46;CD335
Gene Symbols NCR1;LY94
Host Species Mouse
Immunogen Human IgG1-Fc - fused to a recombinant fragment corresponding to 135 amino acids from the ectodomain region of human NKp46.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_001138929
Test Specificity Clone D2-9A5 recognizes the whole NKp46 and NKp46-D2, but does not recognize NKp46-D1. It targets an epitope with in 135 amino acids from the extracellular domain.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.