Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NKTR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115655
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115655 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115655 Supplier Invitrogen™ Supplier No. PA5115655
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NKTR.

This gene encodes a membrane-anchored protein with a hydrophobic amino terminal domain and a cyclophilin-like PPIase domain. It is present on the surface of natural killer cells and facilitates their binding to targets. Its expression is regulated by IL2 activation of the cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NKTR
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Nktr
Gene Accession No. P30414, P30415
Gene Alias 5330401F18Rik; cyclophilin-related protein; D9Wsu172e; natural killer cell triggering receptor; natural killer triggering receptor; natural killer tumor recognition protein; natural killer tumor recognition sequence; natural killer tumor recognition sequence-like; natural killer-tumor recognition sequence; natural-killer cells cyclophilin-related protein; NKTR; NK-TR protein; NK-tumor recognition protein; p104; Peptidyl-prolyl cis-trans isomerase NKTR; PPIase; Rotamase
Gene Symbols Nktr
Host Species Rabbit
Immunogen A synthesized peptide derived from human NKTR(Accession P30414), corresponding to amino acid residues E800-E850.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100364165, 18087, 4820
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.