Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nkx1.1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568927
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568927 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568927 Supplier Invitrogen™ Supplier No. PA568927
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Dog: 82%; Human: 75%; Mouse: 100%; Pig: 75%; Rat: 79%; Yeast: 77%; Zebrafish: 75%.

Nkx1.1 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nkx1.1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Nkx1-1
Gene Accession No. G3UXB3
Gene Alias homeobox protein 153; homeobox protein SAX-2; HPX153; HPX-153 homeobox; HSPX153; NK1 homeobox 1; NK1 transcription factor related, locus 1; NK1 transcription factor related, locus 1 (Drosophila); NK1 transcription factor-related protein 1; Nkx-1.1; NKX1-1; RGD1564874; Sax2; spinal cord axial homeobox gene 2
Gene Symbols Nkx1-1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human Nkx1.1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 672284
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.