Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKX3-2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501733875 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-3875 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-3875 Supplier Proteintech Group Inc Supplier No. 224911AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

BAPX1 is the mammalian homolog of the Drosophila bagpipe homeobox gene and is expressed in the splanchnic mesoderm and embryonic skeleton. It is one of the earliest developmental markers for the sclerotome portion of the somite and the gut mesentery. BAPX1 is required for normal skeletal development; homozygous inactivating mutations in the BAPX1 gene result in spodylo-megaepiphyseal-metaphyseal dysplasia (SMMD). It has also been suggested to play a role in the proper development of the mammalian gut and is required for distal stomach development as part of a BARX1-dependent pathway.

Specifications

Antigen NKX3-2
Applications Western Blot
Classification Polyclonal
Concentration 0.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NKX3-2
Gene Accession No. P78367, P97503
Gene Alias BAPX1, Homeobox protein Nkx 3.2, NK3 homeobox 2, NKX3 2, NKX3.2, NKX3-2, NKX3B
Gene Symbols NKX3-2
Host Species Rabbit
Immunogen NKX3-2 Fusion Protein Ag17581
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12020, 579
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.