Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NLK Antibody, R&D Systems™
SDP

Catalog No. NB432163
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB432163 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB432163 Supplier R&D Systems Supplier No. NBP18604725UL

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

NLK Polyclonal specifically detects NLK in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Simple Western, Western Blot.

Specifications

Antigen NLK
Applications Immunocytochemistry/Immunofluorescence, Simple Western, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 ug/mL, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL
Formulation PBS (pH 7.2) and 40% Glycerol
Gene Alias DKFZp761G1211, EC 2.7.11, EC 2.7.11.24, FLJ21033, LAK1, nemo like kinase, nemo-like kinaseserine/threonine-protein kinase NLK, Protein LAK1
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:LRGPHKQPSLPVLYTLSSQATHEAVHLLCRMLVFDPSKRISAKDALAHPYLDEGRLRYHTCMCKCCFSTSTGRVYTSDFEPVTNPKFDDTFEKNLSSVRQVKEIIHQFILEQQKGNRVPLCINPQSAAFKSFISSTVAQPS
Purification Method Immunogen affinity purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Protein Kinase, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 51701
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.