Learn More
Invitrogen™ NLRP3 Monoclonal Antibody (CL0210)

Description
Immunogen sequence: FKMHLEDYPP QKGCIPLPRG QTEKADHVDL ATLMIDFNGE EKAWAMAVWI FAAINRRDLY EKAKRDEPKW GSDNARVSNP TVICQEDSIE EEWMGLLEYL SRISICKMKK DYRKKYR Highest antigen sequence indentity to the following orthologs: Mouse - 79%, Rat - 78%.
Specifications
Specifications
| Antigen | NLRP3 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | CL0210 |
| Concentration | 1.00 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | NLRP3 |
| Gene Accession No. | Q96P20 |
| Gene Alias | AGTAVPRL; AII; AII/AVP; Angiotensin/vasopressin receptor AII/AVP-like; AVP; C1orf7; caterpiller protein 1.1; Cias1; CLR1.1; cold autoinflammatory syndrome 1 homolog; cold autoinflammatory syndrome 1 protein; cold autoinflammatory syndrome 1 protein homolog; Cold-induced autoinflammatory syndrome 1 protein; cryopyrin; FCAS; FCAS1; FCU; FLJ95925; mast cell maturation inducible protein 1; mast cell maturation-associated-inducible protein 1; Mmig1; MWS; NACHT domain-, leucine-rich repeat-, and PYD-containing protein 3; NACHT, LRR and PYD containing protein 3; NACHT, LRR and PYD domains-containing protein 3; NACHT/LRR/pyrin domain-containing protein 3; NALP3; NLR family pyrin domain containing 3; NLR family, pyrin domain containing 3; Nlrp3; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 3; PYPAF1; PYRIN-containing APAF1-like protein 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.