Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NLRP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115661
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115661 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115661 Supplier Invitrogen™ Supplier No. PA5115661
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NLRP3.

This gene encodes a pyrin-like protein containing a pyrin domain, a nucleotide-binding site domain, and a leucine-rich repeat motif. This protein interacts with the apoptosis-associated speck-like protein PYCARD/ASC, which contains a caspase recruitment domain, and is a member of the NALP3 inflammasome complex. This complex functions as an upstream activator of NF-kappaB signaling, and it plays a role in the regulation of inflammation, the immune response, and apoptosis. Mutations in this gene are associated with familial cold autoinflammatory syndrome, and neonatal-onset multisystem inflammatory disease. Multiple alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene. Alternative 5' UTR structures are suggested by available data; however, insufficient evidence is available to determine if all of the represented 5' UTR splice patterns are biologically valid.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NLRP3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NLRP3
Gene Accession No. D4A523, Q8R4B8, Q96P20
Gene Alias AGTAVPRL; AII; AII/AVP; Angiotensin/vasopressin receptor AII/AVP-like; AVP; C1orf7; caterpiller protein 1.1; Cias1; CLR1.1; cold autoinflammatory syndrome 1 homolog; cold autoinflammatory syndrome 1 protein; cold autoinflammatory syndrome 1 protein homolog; Cold-induced autoinflammatory syndrome 1 protein; cryopyrin; FCAS; FCAS1; FCU; FLJ95925; mast cell maturation inducible protein 1; mast cell maturation-associated-inducible protein 1; Mmig1; MWS; NACHT domain-, leucine-rich repeat-, and PYD-containing protein 3; NACHT, LRR and PYD containing protein 3; NACHT, LRR and PYD domains-containing protein 3; NACHT/LRR/pyrin domain-containing protein 3; NALP3; NLR family pyrin domain containing 3; NLR family, pyrin domain containing 3; Nlrp3; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 3; PYPAF1; PYRIN-containing APAF1-like protein 1
Gene Symbols NLRP3
Host Species Rabbit
Immunogen A synthesized peptide derived from human NLRP3(Accession Q96P20), corresponding to amino acid residues L1005-W1036.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114548, 216799, 287362
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.