Learn More
TARGETMOL CHEMICALS INC NM-3 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T33701
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NM-3 is an orally available anti-angiogenic inhibitor with anti-tumour activity.NM-3 is used as a radiation modulator in vitro and in vivo.NM-3 inhibits vascular endothelial growth factor (VEGF) thereby inhibiting the proliferation of endothelial cells. This inhibits the proliferation of endothelial cells. NM-3 is associated with a mechanism of apoptosis induction by reactive oxygen species. purity: 98%
Catalog No. 50-258-0294
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.