Learn More
Abcam NMDA, Excitotoxic Amino Acid, >99% Purity, Solid, 100 mM/mL
Description
- Molecular Weight: 147.13 Da
- Purity: >99%
- Description: Excitotoxic amino acid; prototypic agonist at the ionotropic NMDA glutamate receptor
- Form: Solid
- CAS Number: 6384-92-5
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Content And Storage | Store at 4°C. |
| Target Molecule | NMDA Receptor |
| Form | Solid |
| Shipping Condition | Ambient |
| Formulation | 100% NMDA |
| Quantity | 100 mM/mL |
| For Use With (Application) | Long-term potentiation, ischemia, epilepsy research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.