Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NMDAR1 Monoclonal Antibody (54.1)
GREENER_CHOICE

Catalog No. 320500
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-050-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-050-0 Supplier Invitrogen™ Supplier No. 320500
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

This antibody specifically recognizes the ~103 kDa NMDAR1 protein. Cross-reactivity with other NMDA receptor proteins has not been observed.

NMDAR1 encodes a protein that is a critical subunit of N-methyl-D-aspartate receptors, members of the glutamate receptor channel superfamily which are heteromeric protein complexes with multiple subunits arranged to form a ligand-gated ion channel. These subunits play a key role in the plasticity of synapses, which is believed to underlie memory and learning. Cell-specific factors are thought to control expression of different isoforms, possibly contributing to the functional diversity of the subunits. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NMDAR1
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 54.1
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene GRIN1
Gene Accession No. P35438, P35439, Q05586
Gene Alias GluN1; Glun1r; GluRdelta1; Glurz1; GluRzeta1; glutamate [NMDA] receptor subunit zeta 1; glutamate [NMDA] receptor subunit zeta-1; glutamate ionotropic receptor NMDA type subunit 1; glutamate receptor; glutamate receptor ionotropic, NMDA 1; glutamate receptor subunit zeta 1; glutamate receptor, ionotropic, NMDA1 (zeta 1); glutamate receptor, ionotropic, N-methyl D-aspartate 1; GRIN1; M100174; MRD8; neurotransmitter receptor; NMDA R1 receptor C1 cassette; NMDA receptor 1; NMDA1; Nmdar; NMDAR1; NMD-R1; NMDZ1; N-methyl-D-aspartate glutamate receptor; N-methyl-D-aspartate receptor 1; N-methyl-D-aspartate receptor channel, subunit zeta-1; N-methyl-D-aspartate receptor subunit NR1; NMZ1; NR1; NR1C1; NR1C2; NR1C2'; NR1N1; pGluN1; Rgsc174; RP11-350O14.1
Gene Symbols GRIN1
Host Species Mouse
Immunogen Fusion protein containing sequence from the intracellular loop between transmembrane regions III and IV of NMDAR1.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14810, 24408, 2902
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.