Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NMDAR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. OPA104011 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OPA104011 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OPA104011 Supplier Invitrogen™ Supplier No. OPA104011
  • $598.00
    $489.65 / Each of 1

    Save $108.35

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

This antibody is predicted to react with canine, chicken, human, non-human primate and Xenopus based on 100% sequence homology. In Western blot, this antibody detects an ~100 kDa protein representing the NR1 N1 splice variant from rat brain homogenate. It shows no cross-reactivity with the NR1 C1, C2, or C2 splice variants. Reconstitute with 50 μL of PBS. After reconstitution the antibody should be aliquotted and stored at -20ºC.

NMDAR1 encodes a protein that is a critical subunit of N-methyl-D-aspartate receptors, members of the glutamate receptor channel superfamily which are heteromeric protein complexes with multiple subunits arranged to form a ligand-gated ion channel. These subunits play a key role in the plasticity of synapses, which is believed to underlie memory and learning. Cell-specific factors are thought to control expression of different isoforms, possibly contributing to the functional diversity of the subunits. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NMDAR1
Applications Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation 5mM Ammonium bicarbonate with no preservative
Gene GRIN1
Gene Accession No. P35438, P35439
Gene Alias GluN1; Glun1r; GluRdelta1; Glurz1; GluRzeta1; glutamate [NMDA] receptor subunit zeta 1; glutamate [NMDA] receptor subunit zeta-1; glutamate ionotropic receptor NMDA type subunit 1; glutamate receptor; glutamate receptor ionotropic, NMDA 1; glutamate receptor subunit zeta 1; glutamate receptor, ionotropic, NMDA1 (zeta 1); glutamate receptor, ionotropic, N-methyl D-aspartate 1; GRIN1; M100174; MRD8; neurotransmitter receptor; NMDA R1 receptor C1 cassette; NMDA receptor 1; NMDA1; Nmdar; NMDAR1; NMD-R1; NMDZ1; N-methyl-D-aspartate glutamate receptor; N-methyl-D-aspartate receptor 1; N-methyl-D-aspartate receptor channel, subunit zeta-1; N-methyl-D-aspartate receptor subunit NR1; NMZ1; NR1; NR1C1; NR1C2; NR1C2'; NR1N1; pGluN1; Rgsc174; RP11-350O14.1
Gene Symbols GRIN1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acid residues specific to the NR1 subunit of rat GRIN1, N1 splice variant insert conjugated to KLH.
Purification Method Affinity Chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14810, 24408
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.