Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NMDAR3A/NMDAR3B Monoclonal Antibody (K35/40.1)
GREENER_CHOICE

Catalog No. 01671886
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-671-886 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-671-886 Supplier Invitrogen™ Supplier No. MA110881
  • $639.00
    $505.00 / Each of 1

    Save $134.00

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-10881 detects Glutamate NMDA Receptor NR3A/B Subunit from human, mouse, and rat samples. This antibody does not cross-react with subunits NR1 or NR2A-D. MA1-10881 has been successfully used in Western blot and immunoprecipitation applications. The MA1-10881 immunogen is Fusion protein of NR3A amino acids 780-914 from the extracellular III-IV loop.

Grin3a is an NMDA (N-methyl-D-aspartate) receptor subtype of glutamate-gated ion channels with reduced single-channel conductance, low calcium permeability and low voltage-dependent sensitivity to magnesium. Grin3a is mediated by glycine and may play a role in the development of dendritic spines. Further, Grin3a may play a role in PPP2CB-NMDAR mediated signaling mechanism. Grin3a belongs to the superfamily of glutamate-regulated ion channels, and function in physiological and pathological processes in the central nervous system. Grin3a shows greater than 90% identity to the corresponding subunit in rat. Studies in the knockout mouse deficient in Grin3a suggest that the protein may be involved in the development of synaptic elements by modulating NMDA receptor activity. Diseases associated with GRIN3A include Lingual-Facial-Buccal Dyskinesia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NMDAR3A/NMDAR3B
Applications Western Blot
Classification Monoclonal
Clone K35/40.1
Concentration 1.4 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene GRIN3B
Gene Accession No. A2AIR5, O60391, Q8TCU5, Q8VHN2, Q91ZU9, Q9R1M7
Gene Alias 6430537F04; A830097C19Rik; chi-1; GluN3A; GluN3B; glutamate [NMDA] receptor subunit 3A; glutamate [NMDA] receptor subunit 3B; glutamate ionotropic receptor NMDA type subunit 3A; glutamate ionotropic receptor NMDA type subunit 3B; glutamate receptor; Glutamate receptor chi-1; Glutamate receptor ionotropic, NMDA 3A; glutamate receptor ionotropic, NMDA 3B; glutamate receptor ionotropic, NMDA3A; glutamate receptor, ionotropic, NMDA3B; glutamate receptor, ionotropic, N-methyl-D-aspartate 3A; glutamate receptor, ionotropic, N-methyl-D-aspartate 3B; GRIN3A; Grin3b; KIAA1973; mKIAA1973; NMDA receptor 4; NMDA receptor subunit 3B; NMDA type glutamate receptor subunit NR3B; NMDAR3A; NMDAR3B; NMDAR-L; NMDAR-L1; N-methyl-D-aspartate receptor; N-methyl-D-aspartate receptor subtype 3A; N-methyl-D-aspartate receptor subtype 3B; N-methyl-D-aspartate receptor subunit NR3B; NR3; NR3A; NR3B; Nr4
Gene Symbols GRIN3A, GRIN3B
Host Species Mouse
Immunogen Fusion protein of NR3A amino acids 780-914 from the extracellular III-IV loop.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116443, 116444, 170483, 170796, 191573, 242443
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.