Learn More
TARGETMOL CHEMICALS INC NMDI14 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1223510MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NMDI14 is an inhibitor of nonsense mediated RNA decay (NMD). purity: 99%
Catalog No. 50-257-7429