Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NME1 Monoclonal Antibody (13C1)
GREENER_CHOICE

Catalog No. PIMA534938
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534938 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534938 Supplier Invitrogen™ Supplier No. MA534938
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive Control: K562 cell, A549 cell, HepG2 cell, 293 cell, human liver carcinoma tissue, human thyroid tissue, human skin tissue, human breast carcinoma tissue.

NME1 was identified because of its reduced mRNA transcript levels in highly metastatic cells. Nucleoside diphosphate kinase (NDK) exists as a hexamer composed of 'A' (encoded by this gene) and 'B' (encoded by NME2) isoforms. Mutations in the gene have been identified in aggressive neuroblastomas. Two transcript variants encoding different isoforms have been found for this gene. Co-transcription of this gene and the neighboring downstream gene (NME2) generates naturally-occurring transcripts (NME1-NME2), which encodes a fusion protein comprised of sequence sharing identity with each individual gene product.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NME1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 13C1
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4
Gene NME1
Gene Accession No. P15531
Gene Alias AL024257; AWD; expressed in non-metastatic cells 1; expressed in non-metastatic cells 1 protein; expressed in non-metastatic cells 1 protein (NM23A) (nucleoside diphosphate kinase); expressed in non-metastatic cells 1, protein; expressed in non-metastatic cells 1, protein (NM23A) (nucleoside diphosphate kinase); GAAD; Granzyme A activated DNase (GAAD); Granzyme A-activated DNase; GZMA activated DNase; included; metastasis inhibition factor NM23; NB; NBR-A; NBR-B; NBS; NDK A; NDK A 1; NDK A 2; NDK NBR-A; NDK NBR-B; NDKA; NDP kinase A; NDP kinase A 1; NDP kinase A 2; NDP kinase beta; NDP kinase NBR-A; NDPKA; NDPK-A; NM23; NM23A; NM23-C1; NM23-H1; NM23H1B; nm23-M1; NME/NM23 nucleoside diphosphate kinase 1; Nme1; NME1-1; NME1-NME2 spliced read-through transcript; non-metastatic cells 1, protein (NM23A) expressed in; nucleoside diphosphate kinase A; nucleoside diphosphate kinase A 1; nucleoside diphosphate kinase A 2; nucleoside diphosphate kinase NBR-B; nucleoside-diphosphate kinase 1; nucleoside-diphosphate kinase NBR-A; nucleotide diphosphate kinase; nucloside diphosphate kinase; tumor metastatic process-associated protein
Gene Symbols NME1
Host Species Mouse
Immunogen Synthetic peptide within Human NM23 aa 1-50.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4830
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.