Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NNC 26-9100
GREENER_CHOICESDP

Catalog No. 244010
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-401-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 24-401-0 Supplier Tocris Bioscience™ Supplier No. 2440/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective sst4 agonist

NNC 26-9100 is a somatostatin sst4 receptor agonist that displays > 100-fold selectivity over sst2 receptors (Ki values are 6 and 621 nM for sst4 and sst2 receptors respectively). Potently inhibits forskolin -induced cAMP accumulation (EC50 = 26 nM).

Chemical Identifiers

CAS 199522-35-5
Molecular Formula C22H25BrCl2N6S

Specifications

Quantity 10 mg
Formula Weight 556.35
Purity >99%
Molecular Weight (g/mol) 556.348
Chemical Name or Material NNC 26-9100
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.