Learn More
Abcam NNGH, Matrix Metalloprotease Inhibitor, >98% Purity, Solid, 5 mg
Description
- Molecular Weight: 316.38 Da
- Purity: >98%
- Form: Solid
- Inhibitor Activity: MMP3 (Stromelysin-1) inhibitor; shows broad spectrum inhibition activity at other MMPs
- Characteristics: Cell permeable
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Content And Storage | Store at ambient temperature. |
| Target Molecule | Matrix Metalloprotease |
| Substrate Properties | Chemical Substrate |
| Buffer | 100% NNGH |
| Form | Solid |
| Shipping Condition | Ambient |
| Formulation | 100% NNGH |
| Quantity | 5 mg |
| For Use With (Application) | Matrix metalloprotease inhibition |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.