Learn More
ProteinSimple No secondary Detection Module
Supplier:  ProteinSimple DM003
Encompass
Compatible with Jess, Abby, Wes, Peggy Sue and Sally Sue systems. Module includes: Luminol-S, Peroxide, Antibody Diluent 2, Streptavidin-HRP.
Catalog No. NC1208999