Learn More
                                    Paul N Gardner Company NO5 ZAHN CUP
Supplier: Paul N Gardner Company VI2105
            Calibrated, but ** NOT CERTIFED ** S90 ,ZAHN Signature Series Dip Viscosity Cup, No. 5 S90/Zahn Viscosity Cup,Seconds Range:10 to 60,Centistoke Range:219 to 1627,Cup flow characteristics are defined by a mathematical formula and standard (G) series oils. ** For calibrated and certified cup please see Fisher Scientific part # NC0593533 **
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.