Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NO66 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564895
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564895 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564895 Supplier Invitrogen™ Supplier No. PA564895
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RVVETSALLCT AQHLAAVQSS GAPATASGPQ VDNTGGEPAW DSPLRRVLAE LNRIPSSRRR AARLFEWLIA PMPPDHFYRR LWEREAVLVR RQDHTYYQGL FSTADLDSML RNEEVQFGQH LDAARYINGR R Highest antigen sequence identity to the following orthologs - mouse 61%, rat 66%.

Oxygenase that can act as both a histone lysine demethylase and a ribosomal histidine hydroxylase. Specifically demethylates 'Lys-4' (H3K4me) and 'Lys-36' (H3K36me) of histone H3, thereby playing a central role in histone code. Preferentially demethylates trimethylated H3 'Lys-4' (H3K4me3) and monomethylated H3 'Lys-4' (H3K4me1) residues, while it has weaker activity for dimethylated H3 'Lys-36' (H3K36me2). Also catalyzes the hydroxylation of 60S ribosomal protein L8 on 'His-216'. Acts as a regulator of osteoblast differentiation via its interaction with SP7/OSX by demethylating H3K4me and H3K36me, thereby inhibiting SP7/OSX-mediated promoter activation. May also play a role in ribosome biogenesis and in the replication or remodeling of certain heterochromatic region. Participates in MYC-induced transcriptional activation. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen NO66
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RIOX1
Gene Accession No. Q9H6W3
Gene Alias 2410016O06Rik; 60S ribosomal protein L8 histidine hydroxylase; bifunctional lysine-specific demethylase and histidyl-hydroxylase NO66; C14orf169; chromosome 14 open reading frame 169; FLJ21802; histone lysine demethylase NO66; hsNO66; lysine-specific demethylase NO66; Mapjd; MNCb-7109; myc associated protein with JmjC domain; MYC-associated protein with JmjC domain; NO66; nucleolar protein 66; RGD1307704; Ribosomal oxygenase 1; Ribosomal oxygenase NO66; RIKEN cDNA 2410016O06 gene; Riox1; ROX; similar to RIKEN cDNA 2410016O06; up-regulated in lung cancer 2; URLC2
Gene Symbols RIOX1
Host Species Rabbit
Immunogen Recombinant Human NO66. Recombinant protein control fragment (Product #RP-105694).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79697
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.