Learn More
Noble Agar, Ultrapure
Description
Gelling agent essentially free of impurities. Used mainly in biochemistry and molecular biology procedures wherever increased purity is required.
Typical Agar Preparation:
- Plates: 15 g/L of media; 30-35 ml per 90 mm plate
- Top Agar: 7 g/L of media
Autoclave for 20 minutes and swirl gently to evenly distribute the agar. If antibiotics are to be added, allow the agar to cool to 50°C. When pouring plates, gently swirl the medium. A Bunsen burner flame can be passed over the surface of the plates to remove any air bubbles. After the medium has hardened, invert the plates and store at 4°C.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Format: Pale cream powder
- CAS: 9002-18-0
- MDL Number: MFCD00081288
Typical Agar Preparation
Plates: 15g/L of media; 30 to 35mL per 90mm plate
Top Agar: 7g/L of media
Autoclave for 20 minutes and swirl gently to evenly distribute the agar. If antibiotics are to be added, allow the agar to cool to 50°C. When pouring plates, gently swirl the medium. A Bunsen burner flame can be passed over the surface of the plates to remove any air bubbles. After the medium has hardened, invert the plates and store at 4°C
Specifications
Specifications
| Color | Pale cream |
| Form | Powder |
| Grade | Ultrapure |
| Media Type | Noble Agar |
| Product Type | Nutrient Agar |
| For Use With (Application) | Used as a Gelling Agent in Biochemistry and Molecular Biology Procedures Where Increased Purity is Required. |
| Packaging Type | Plastic Bottle |
| Quantity | 1 kg |
Safety and Handling
| EINECSNumber | 232-658-1 |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.