Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOBOX Monoclonal Antibody (OTI8C7), TrueMAB™, OriGene
SDP

Catalog No. 501682141
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-2141 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-2141 Supplier OriGene Supplier No. TA808449
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NOBOX is a transcription factor which may play a role in oogenesis. It binds preferentially to the DNA sequences 5′-TAATTG-3′, 5′-TAGTTG-3′ and 5′-TAATTA-3′.

Specifications

Antigen NOBOX
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI8C7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene NOBOX
Gene Accession No. O60393
Gene Alias OG-2, OG2, OG2X, POF5, TCAG_12042
Gene Symbols NOBOX
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1-271 of human NOBOX produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 135935
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.