Learn More
Cole-Parmer Advantec 800700 Nobuto Blood Filter Strip, 100/pk
Supplier:  Cole-Parmer 0664440
Encompass_Preferred
These filter strips are ideal for collection of applications. Collect blood on the narrow end of the strip and place on a drying rack for easy and efficient sample handling. Each strip can hold approximately 0.1 mL of blood or 0.04 mL of serum. The strips are produced with high-purity cellulose paper with a fine, textile like structure, which is ideal for uniform sampling.
Catalog No. NC9007301
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.