Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOC2L Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01675610
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-610 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-675-610 Supplier Invitrogen™ Supplier No. PA141264
  • $587.00
    $487.70 / Each of 1

    Save $99.30

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Suggested positive control: Hela whole cell extract, HeLa cell lysate. Reacts with Chimpanzee and Rhesus Monkey. Predicted to react with Chicken, Mouse and Rat.

INHAT (inhibitor of acetyltransferases) complex binds to histones and masks them from being substrate for acetyltransferase. Endogenous INHAT subunits, which include the Set/TAF-Ibeta oncoprotein, associate with chromatin in vivo and can block coactivator mediated transcription when transfected in cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOC2L
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 100μg/mL BSA and 0.02% sodium azide
Gene NOC2L
Gene Accession No. Q3SYU1, Q9Y3T9
Gene Alias AA410003; AF155546; DKFZp564C186; FLJ35172; hypothetical protein LOC508638; NET15; NET7; NIR; NOC2 like nucleolar associated transcriptional repressor; NOC2L; NOC2L nucleolar associated transcriptional repressor; NOC2-like; NOC2-like nucleolar associated transcriptional repressor; NOC2-like protein; novel INHAT (inhibitor of histone acetyltransferase) repressor; novel INHAT repressor; nucleolar complex associated 2 homolog; nucleolar complex associated 2 homolog (S. cerevisiae); nucleolar complex protein 2 homolog; PPP1R112; Protein NOC2 homolog; protein phosphatase 1, regulatory subunit 12; RGD1309387
Gene Symbols NOC2L
Host Species Rabbit
Immunogen Synthetic peptides of amino acids 87-103 and 231-246 of human INHAT-2.
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26155, 508638
Target Species Human, Monkey, Bovine
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.