Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Nocardamine, Deferrioxamine E, Antibiotic Agent, 1 mg

Catalog No. 502705484
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5484 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5484 Supplier Abcam Supplier No. AB1448421MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Siderophore iron chelator that induces morphological changes in insect cells.

  • Antibiotic agent
  • Siderophore iron chelator
  • Induces morphological changes in insect cells
  • Shows weak antimicrobial against Gram-positive bacteria
  • Purity >95%
  • MW 600.7 Da

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Target Molecule Iron
Buffer 100% Nocardamine
Content And Storage Store at -20°C.
Substrate Properties Chemical Substrate
Formulation 100% Nocardamine
Quantity 1 mg
For Use With (Application) Antibiotic agent, Iron chelation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.