Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Nociceptin (1-7), ORL1 Receptor Agonist, Solid, 1 mg

Catalog No. 502707314
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7314 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7314 Supplier Abcam Supplier No. AB1207411MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bioactive fragment of nociceptin that acts as an ORL1 (NOP) receptor agonist.

  • Molecular weight: 655.71 g/mol
  • Purity: >95%
  • Form: Solid
  • Activity: ORL1 (NOP) receptor agonist; reduces nociception induced hyperalgesia and displays antinociceptive activity
  • Storage: Store at -20°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule ORL1 Receptor
Shipping Condition Ambient
Content And Storage Store at -20°C.
Form Solid
Quantity 1 mg
For Use With (Application) ORL1 (NOP) receptor agonist
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.