Learn More
Description
PA3-204 detects Nociceptin in human, rat and mouse samples. PA3-204 has successfully been used in immuncytochemical and immunohistochemical (paraffin) procedures. The PA3-204 immunogen is a synthetic peptide corresponding to the amino acids FGGFTGARKSARKLANQ of Nociceptin.
Specifications
Specifications
| Antigen | Nociceptin |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | whole serum with 0.02% sodium azide |
| Gene | PNOC |
| Gene Accession No. | Q13519, Q62923, Q64387 |
| Gene Alias | N/OFQ; N23K; N23K/N27K; neuropeptide nociceptin 1; nociceptin; Nocistatin; NOP; Npnc1; OFQ; OFQ Orphanin FG; OFQ/N; ORL1 receptor agonist; Orphanin FQ; Orphanin FQ2; Pnoc; ppN/OFQ; PPNOC; pre-pro-N/OFQ; prepronociceptin; Prepronociceptin (neuropeptide nociceptin) (N23K); pronociceptin; proorphanin |
| Gene Symbols | PNOC |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
