Learn More
Nocodazole, MP Biomedicals™
Catalog No. MP215240510 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
Catalog No. Quantity
MP215240510 10 mg
1 options
Catalog No. MP215240510 Supplier MP Biomedicals, Inc Supplier No. 0215240510
Add to Cart
Add to Cart
Used in Cell Biology to synchronize the cell division cycle, as well as in cell studies and cancer research when trying to understand the microspindles in the cell replication process.
  • Used in cell division & replication research, cancer research, microscopy, and flow cytometry
  • Nocodazole is usually used at a concentration of 40–100 ng/mL of culture medium for a duration of 12 to 18 hours
  • Prolonged arrest of cells in mitosis due to nocodazole treatment typically results in cell death by apoptosis
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10 mg