Learn More
STEMCELL Technologies Nocodazole, Size: 10 mg
SDP
Supplier:  STEMCELL Technologies 74072
Encompass
Nocodazole is an anti-mitotic agent that reversibly interferes with the polymerization of microtubules (De Brabander et al.). Nocodazole binds to beta-tubulin and disrupts microtubule assembly/disassembly dynamics, impairing formation of the metaphase spindles during the cell division cycle. This prevents mitosis by inducing a G2/M-phase arrest and induces apoptosis in tumor cells (Jordan et al.).GENOME EDITING· Synchronizes human pluripotent stem cells (hPSCs) in the G2/M cell cycle phase, thereby increasing the efficiency of homology-directed repair (HDR) in CRISPR-Cas9 genome editing (Lin et al.; Yang et al.). CANCER RESEARCH· Induces apoptosis in leukemic cells without harming T cells and/or mesenchymal stromal cells that were recovered from the same patients (Frezzato et al.).
Catalog No. 50-197-6175
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.