Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nod2 Monoclonal Antibody (26mNOD2), eBioscience™, Invitrogen™

Catalog No. 5013221
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-132-21 50 μg
50-132-22 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-132-21 Supplier Invitrogen™ Supplier No. 14585881
  • $143.00
    $107.65 / Each of 1

    Save $35.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

Description: The monoclonal antibody 26mNOD2 reacts with mouse NOD2. NOD2, also referred to as caspase recruitment domain-containing protein 15 (CARD 15), is a cytosolic protein of approximately 115kDa. NOD2 contains a central NOD (nucleotide oligomerization domain) that is flanked by a C-terminal leucine-rich repeat (LRR) domain and an N-terminal CARD domain. The LRR confers recognition of microbial components such as muramyl dipeptide and the CARD domain confers the ability to interact with RIP2 via homotypic interaction with the RIP2 CARD. This interaction leads to downstream activation of NF-kappaB. The antibody 26mNOD2 has been shown to recognize protein from transgenic and transfected cells. Refer to references for expression in endogenous cells. Applications Reported: This 26mNOD2 antibody has been reported for use in immunoblotting (WB). Applications Tested: This 26mNOD2 antibody has been tested by immunoblotting of lysates of 293T cells transfected with mouse NOD2. This antibody has been validated on transfected cell lines and cells from NOD2-transgenic mice. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

NOD2 is a member of the apoptosis regulating protein family that includes caspase recruitment-domains, as well as Apaf-1 and NOD1. It contains two N-terminal CARDs, a nucleotide binding domain (NBD), and multiple C-terminal leucine-rich repeats (LRRs). NOD2 is expressed in monocytes (whereas NOD1 is expressed in multiple tissues). NOD2 plays a role in regulating NF-kappaB, acts as an intracellular receptor for bacterial lipopolysaccharides, and contributes to inflammatory bowel disease (IBD) and Crohn's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nod2
Applications Western Blot
Classification Monoclonal
Clone 26mNOD2
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene NOD2
Gene Accession No. Q8K3Z0
Gene Alias ACUG; BLAU; CARD15; caspase recruitment domain family, member 15; caspase recruitment domain protein 15; caspase recruitment domain-containing protein 15; CD; CLR16.3; F830032C23Rik; IBD1; Inflammatory bowel disease protein 1; NLR family, CARD domain containing 2; Nlrc2; NOD 2; NOD2; NOD-2; NOD2 protein; NOD2B; NOD-like receptor C2; nucleotide binding oligomerization domain containing 2; nucleotide-binding oligomerization domain 2; nucleotide-binding oligomerization domain containing 2; nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 2; nucleotide-binding oligomerization domain-containing protein 2; PSORAS1
Gene Symbols NOD2
Host Species Rat
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 257632
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.