Learn More
Enzo Life Sciences Nodularin (100µg). CAS: 118399-22-7
SDP
Supplier:  Enzo Life Sciences ALX350061C100
Encompass_Preferred
Inhibitor of protein phosphatase 1 (PP1) (IC50=1.8nM), protein phosphatase 2A (PP2A) (IC50=0.026nM) and to a lesser extent protein phosphatase 2B (PP2B) (IC50=8.7µm). Similar to microcystin-LR (ALX-350-012) but with increased water solubility. Genotoxic. Hepatoxic. Induces oxidative DNA damage by oxidation of purines. Induces apoptosis in HepG2 cells. Purity: ≥95% (HPLC). Solubility: Soluble in methanol:water (1:1). Long Term Storage: -20°C.
Catalog No. 50-200-8710
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.