Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nogo-A/B Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01675776
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-776 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-675-776 Supplier Invitrogen™ Supplier No. PA141220
  • $586.00
    $477.30 / Each of 1

    Save $108.70

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Suggested positive control: human, mouse or rat brain.

Nogo-B is a novel myelin-associated, intermediate-length splice variant of Nogo belonging to the reticulon family. It is a multidomain neurite growth inhibitory protein containing several discrete regions with growth inhibitory functions and is a component of the CNS myelin that prevents axonal regeneration in the adult vertebrate CNS. Nogo-B along with NGBR mediated chemotaxis in HUVECs and induced tube formation in 3-dimensional cultures. It is mostly found in intact blood vessels, smooth muscle cells, and endothelial cell, with its N terminus oriented extracellularly and is a regulator of vascular homeostasis and remodeling. It plays an important role in sporadic and familial amyotrophic lateral sclerosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nogo-A/B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene RTN4
Gene Accession No. Q99P72, Q9JK11, Q9NQC3
Gene Alias 1110020G17Rik; AA407876; AA409940; AA960376; ASY; C130026I10Rik; Foocen; glut4 vesicle 20 kDa protein; GLUT4 vesicle 20kDa protein; Human NogoA; KIAA0886; MGC116054; mKIAA0886; mKIAA4153; My043; My043 protein; Nbla00271; Nbla10545; neurite growth inhibitor 220; neurite outgrowth inhibitor; Neuroendocrine-specific protein; Neuroendocrine-specific protein C homolog; NgA; NI220/250; NI-220/250; NI-250; NOGO; nogo protein; Nogo-A; Nogo-B; NOGOC; Nogo-C; NSP; NSP-CL; r; rat N; rat NogoA; reticulon 4; reticulon 5; reticulon-4; Reticulon-5; Rtn4; RTN4-A; RTN4-B1; RTN4-B2; RTN4-C; RTN-x; SP1507; Vp20
Gene Symbols RTN4
Host Species Rabbit
Immunogen A synthetic peptide to an internal portion of human Nogo A (between amino acids 1-50).
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100170399, 100338467, 57142, 68585, 83765
Target Species Human, Mouse, Sheep, Rabbit, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.