Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bal Supply N99 Particulate Respirators
Shop All Tip Industries Inc. Products
Click to view available options
With Exhalation Valve:
Yes
Description
- Conforms to NIOSH 42 CFR84
- Protects against solid and liquid aerosols
- Flexible metal nose clip seal with cushioning foam for comfort
- Made of soft, lightweight material with ergonomic shape and stapled straps
Specifications
Specifications
| Aerosol Type | N - Not Resistant to Oil |
| Certifications (NIOSH) | NIOSH 42 CFR 84, N99, approval number: TC-84A-3890 |
| Design Type | Cup/Molded |
| Disposable | Single-use |
| Filter Type | N99 |
| Product Type | General Purpose Particulate Respirator |
| Size | Standard |
| With Exhalation Valve | Yes |
| Filter Efficiency | 99% |
| Latex Free | Yes |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction