Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOL11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103983
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA5103983 100 μL
1 options

Catalog No. PIPA5103983

Supplier: Invitrogen™ PA5103983

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NOL11.

NOL11 is a ribosome biogenesis factor. It may be required for both optimal rDNA transcription and small subunit (SSU) pre-rRNA processing at sites A', A0, 1 and 2b.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOL11
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NOL11
Gene Accession No. Q8BJW5, Q9H8H0
Gene Alias 1500002M01Rik; AU015220; AU019874; L14; Nol11; Nucleolar protein 11
Gene Symbols NOL11
Host Species Rabbit
Immunogen A synthesized peptide derived from human NOL11(Accession Q9H8H0), corresponding to amino acid residues E541-I591.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25926, 688885, 68979
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.