Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nomega-Nitro-L-arginine 98.0+%, TCI America™
SDP

Catalog No. N066025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N066025G 25 g
N06605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N066025G Supplier TCI America Supplier No. N066025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2149-70-4
Molecular Formula C6H13N5O4
Molecular Weight (g/mol) 219.201
MDL Number MFCD00007033
InChI Key MRAUNPAHJZDYCK-BYPYZUCNSA-N
Synonym h-arg no2-oh, ng-nitro-l-arginine, l-nna, nitroarginine, n-nitro-l-arginine, nomega-nitro-l-arginine, n g-nitro-l-arginine, ng-no2-l-arg, n-omega-nitro-l-arginine, n g-nitroarginine
PubChem CID 440005
ChEBI CHEBI:27960
SMILES C(CC(C(=O)O)N)CN=C(N)N[N+](=O)[O-]

Specifications

Color White
Quantity 25 g
Formula Weight 219.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nomega-Nitro-L-arginine
RTECSNumber RM2982100
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.