Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Emergency Fire Blanket
SCI_ED

Lightweight and easy to apply.

Supplier:  Sellstrom Manufacturing 97450/EMD

Catalog No. S47559B


Description

Description

  • Smothers clothing fires and other small fires
  • Made of fiberglass and hemmed on all four sides with Kevlar™ thread
  • Brass grommets on corners
  • Machine washable
  • 1.5 x 1.8m (5 x 6 ft.)
  • Red, heavy-duty vinyl pouch protects blanket from exposure to UV rays, chemicals, moisture dust, salt air, insects, and temperature extremes
  • Use for rescue operations in offices, industries and hospitals: either to cover the fire, cutting off the oxygen supply or to wrap around a person whose clothes are on fire
Specifications

Specifications

Fire Blanket
Fiberglass
Rescue operations in offices, industries and hospitals
2 lb.
White
72 x 60 in. (182 x 152.4 cm)
Wall hanging Vinyl pouch with pull tab
0.9 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.