Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Nomex™ / Lenzing FR Full Face Hood

Catalog No. 19100456 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Design Type:
Straight Cut Design
Tri-Cut Design
Color:
Black
White
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Design Type Color
19-100-456 Straight Cut Design -
19-100-453 Tri-Cut Design White
19-100-454 Straight Cut Design White
19-100-455 Tri-Cut Design Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-100-456 Supplier PIP™ Supplier No. 9062080NOL7BLKB
  • $98.00
    $76.65 / Each of 1

    Save $21.35

    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

PIP™ Nomex™ / Lenzing FR Full Face Hood features a fabric combination that provides comfort and breathability

  • Provides comfort and breathability

Specifications

Product Type Fire Protectant Hood
Size Universal
Closure Type Pull-over
Disposable Reusable
Design Type Straight Cut Design
Quantity 1 Ea.
Includes Hood Only
For Use With (Application) Heat/Fire Applications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.