Learn More
Bachem (Glu13·17·20)-Osteocalcin (1-46) (mouse) Trifluoroacetate
SDP
Supplier:  Bachem 4057685.1000
Encompass_Preferred
(Glu13·17·20)-Osteocalcin (1-46) (mouse) Trifluoroacetate, H-Tyr-Leu-Gly-Ala-Ser-Val-Pro-Ser-Pro-Asp-Pro-Leu-Glu-Pro-Thr-Arg-Glu-Gln-Cys-Glu-Leu-Asn-Pro-Ala-Cys-Asp-Glu-Leu-Ser-Asp-Gln-Tyr-Gly-Leu-Lys-Thr-Ala-Tyr-Lys-Arg-Ile-Tyr-Gly-Ile-Thr-Ile-OH, 1802086-27-6, C226H351N57O74S2, Mr 5114.75, 1mg. Non-carboxylated osteocalcin was shown to be a bone-specific hormone involved in the regulation of glucose metabolism, for which the Gla peptide acts as precursor and transport form.
Catalog No. 50-194-6910
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.