Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynalon™ Non-Hazardous-Waste Lab Disposal Box
SCI_ED

Catalog No. s29152 Shop All Dynalab Corporation Products
Change view
Click to view available options
Length (English):
12 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
S29152 12 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S29152 Supplier Dynalon™ Supplier No. 797055/EMD
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Easy identification

Non-Hazardous Disposal Box features colorful graphics for quick identification.

  • Assembly instructions on each box
  • Tough 2 mil polyethylene liner
  • Folding design for shipping and storage
  • Closure assures a safe, hygienic disposal
  • Supplied in a case of 6 boxes

Specifications

Closure Type Lid
Description Non-Hazardous Disposal Box
Color Green, White
Dimensions (L x W x H) 12 x 12 x 27 in.
Height (Metric) 685.8 mm
Length (Metric) 304.8 mm
Opening Size (Metric) 177.8 mm
Shape Rectangle
Width (Metric) 304.8 mm
Autoclavable Not Autoclavable
Diameter (English) Opening 7 in.
Diameter (Metric) Opening 177.8 mm
Includes Polyethylene Liner
Liner Material Polyethylene
For Use With (Application) Non-Hazardous Waste
Width (English) 12 in.
Length (English) 12 in.
Height (English) 27 in.
Product Type Disposal Box
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.