Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Non-human Primate IFN-alpha ELISA Kit
SDP

Catalog No. NB120294 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit (1 Plate)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120294 1 Kit (1 Plate)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB120294 Supplier Novus Biologicals™ Supplier No. NBP311725
Only null left
Add to Cart
Add to Cart

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

Specifications

Assay Range 5 - 4000 pg/mL
Assay Sensitivity 2 pg/mL
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Plasma, Serum
For Use With (Application) ELISA
Gene ID (Entrez) 3440
Gene Symbol IFNA2
Quantity 1 Kit (1 Plate)
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym IFNA, IFNA2, IFNA2b, IFN-alphaA, INFA2, interferon alpha 2
Target Non-human Primate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.