Learn More
CritiCore Protective Wear Non-Linting Sandwash Scrub Pants in Navy
Catalog No. 17012844
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-012-844 X-Large
17-012-840 X-Small
17-012-841 Small
17-012-842 Medium
17-012-843 Large
17-012-845 2X-Large
17-012-846 3X-Large
17-012-847 4X-Large
17-012-848 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 17-012-844 Supplier CritiCore Protective Wear Supplier No. 13PA492SWS601XLNVY
Add to Cart
Add to Cart
This item is not returnable. View return policy
These non-linting sandwash scrub pants in navy are soft, moisture-wicking, and breathable, an ideal cleanroom base layer.
  • Navy sandwash
  • Drawstring / elastic waist
  • Polyester cuff leg closure
  • Primary application: Cleanroom Innerwear
Specifications
Size X-Large
Gender Unisex
Product Type Scrub Suit Pants
Material Polyester
Color Navy
Seam Type Stitched
No. of Pockets None