Learn More
ATCC Non-Small Cell Lung Cancer p53 Hotspot Mutation Cell Line Panel
Supplier:  ATCC TCP2030
Encompass
The Non-Small Cell Lung Cancer p53 Hotspot Mutation Cell Panel (ATCC TCP-2030) is comprised of 6 select cell lines derived from the lung that have been sequenced and validated by ATCC. The panel includes both WT p53 cell lines as well as cultures with p53 hotspot mutations at codons 245, 248, or 273.
Catalog No. NC2235212
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.