Learn More
Fisherbrand™ Round-Bottom Polypropylene Test Tubes Without Cap
Catalog No. p-7203039 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
5 mL
14 mL
Sterility:
Non-sterile
Sterile
Graduated:
No
Yes
Packaging:
Bag
Blister Pack
Blister Pk.
Bulk
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Sterility Graduated Packaging
FB1495916 5 mL Non-sterile No Bag
FB1495916A 5 mL Non-sterile No Bulk
FB149567E 14 mL Non-sterile No Bulk
FB149561B 5 mL Sterile No Blister Pk.
FB149561G 14 mL Sterile No Blister Pk.
FB149569A 14 mL Sterile Yes Blister Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. FB1495916
Add to Cart
Add to Cart
This item is not returnable. View return policy
Fisherbrand™'s round bottom tubes provide reliable containment for various laboratory fluid samples and preparations.
  • Ideal for use in laboratory protocols requiring round bottom tubes
  • Ensures reliable handling of liquid samples
  • Polypropylene tubes are best suited for applications requiring greater thermal and chemical stability, while polystyrene tubes offer high optical clarity
  • Available with and without a snap cap - dual-position snap caps offer both vented and fully closed positions.
  • 3,000 RCF rating for polypropylene tubes, 1,400 RCF rating for polystyrene tubes
  • Dimensions and volumes are approximate
TRUSTED_SUSTAINABILITY
Specifications
Color Translucent
Capacity (Metric) 5 mL
Length (Metric) 75 mm
Material Polypropylene
Disposable Single-use
Packaging Bag
Sterility Non-sterile
Product Line Fisherbrand
Graduated No
Closure Type Not Included
Product Type Test Tube