Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nonadecafluorodecanenitrile 98.0+%, TCI America™
SDP

Catalog No. N10955G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N10955G 5 g
N10951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N10955G Supplier TCI America Supplier No. N10955G

Chemical Identifiers

CAS 379215-40-4
Molecular Formula C10F19N
Molecular Weight (g/mol) 495.087
MDL Number MFCD00045610
InChI Key QXBUHLMUCFBZOL-UHFFFAOYSA-N
Synonym Perfluorodecanonitrile, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Nonadecafluorodecanenitrile
PubChem CID 2776067
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluorodecanenitrile
SMILES C(#N)C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Color Yellow
Quantity 5 g
Formula Weight 495.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Nonadecafluorodecanenitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.