Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nonafluoro-1-butanesulfonic Acid 98.0+%, TCI America™
SDP

Catalog No. N070925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N070925G 25 g
N07095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N070925G Supplier TCI America Supplier No. N070925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 375-73-5
Molecular Formula C4HF9O3S
Molecular Weight (g/mol) 300.095
MDL Number MFCD01320794
InChI Key JGTNAGYHADQMCM-UHFFFAOYSA-N
Synonym Perfluoro-1-butanesulfonic Acid
PubChem CID 67815
IUPAC Name 1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulfonic acid
SMILES C(C(C(F)(F)S(=O)(=O)O)(F)F)(C(F)(F)F)(F)F

Specifications

Color Yellow
Boiling Point 212°C
Quantity 25 g
UN Number 2586
Formula Weight 300.09
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Nonafluoro-1-butanesulfonic Acid
RTECSNumber EK5930000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.