Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nonafluoro-1-iodobutane, 97%

Catalog No. AAA1667114
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1667114 25 g
AAA1667106 5 g
AAA1667122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1667114 Supplier Thermo Scientific Chemicals Supplier No. A1667114
Add to Cart

CAS: 423-39-2 | C4F9I | 345.934 g/mol

Nonafluoro-1-iodobutane is used as iodine atom donor in the synthesis of iodocyclohexane. It is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nonafluoro-1-iodobutane is used as iodine atom donor in the synthesis of iodocyclohexane. It is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Not miscible in water.

Notes
Light Sensitive. Store in the dark away from oxidizing agents. Incompatible with alkali metals and light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 423-39-2
Molecular Formula C4F9I
Molecular Weight (g/mol) 345.934
MDL Number MFCD00001062
InChI Key PGRFXXCKHGIFSV-UHFFFAOYSA-N
Synonym perfluorobutyl iodide, nonafluoro-1-iodobutane, nonafluorobutyl iodide, perfluorobutyliodide, n-nonafluorobutyl iodide, perfluoro-n-butyl iodide, 1-iodoperfluorobutane, ccris 9008, 1-iodononafluorobutane, butane, 1,1,1,2,2,3,3,4,4-nonafluoro-4-iodo
PubChem CID 67917
IUPAC Name 1,1,1,2,2,3,3,4,4-nonafluoro-4-iodobutane
SMILES C(C(C(F)(F)I)(F)F)(C(F)(F)F)(F)F

Specifications

Melting Point -89°C to -87°C
Density 2.01
Boiling Point 67°C
Odor Pungent
Refractive Index 1.329
Quantity 25 g
UN Number UN3082
Beilstein 1777546
Sensitivity Light sensitive
Solubility Information Not miscible in water.
Formula Weight 345.93
Percent Purity 97%
Chemical Name or Material Nonafluoro-1-iodobutane
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 207-025-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.