Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nonafluorobutyl Iodide 98.0+%, TCI America™
SDP

Catalog No. N0499100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0499100G 100 g
N049925G 25 g
N0499500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N0499100G Supplier TCI America Supplier No. N0499100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 423-39-2
Molecular Formula C4F9I
Molecular Weight (g/mol) 345.934
MDL Number MFCD00001062
InChI Key PGRFXXCKHGIFSV-UHFFFAOYSA-N
Synonym perfluorobutyl iodide, nonafluoro-1-iodobutane, nonafluorobutyl iodide, perfluorobutyliodide, n-nonafluorobutyl iodide, perfluoro-n-butyl iodide, 1-iodoperfluorobutane, ccris 9008, 1-iodononafluorobutane, butane, 1,1,1,2,2,3,3,4,4-nonafluoro-4-iodo
PubChem CID 67917
IUPAC Name 1,1,1,2,2,3,3,4,4-nonafluoro-4-iodobutane
SMILES C(C(C(F)(F)I)(F)F)(C(F)(F)F)(F)F

Specifications

Color Red-Yellow
Boiling Point 67°C
Quantity 100 g
Formula Weight 345.93
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Nonafluorobutyl Iodide
RTECSNumber EK5360000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.