Learn More
Sigma Organic Chemistry NonafluoropentanoY chlo | 250mg | MFCD00155890
Supplier: Sigma Organic Chemistry CDS002982250MG

NonafluoropentanoY chlo , 250mg
About this Item:
MDL #: MFCD00155890
Molecular Weight: 282.49
UNSPSC Code: 12352200
Chemical Formula: C5ClF9O
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.