Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nonanal, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019974 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9974 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9974 Supplier MilliporeSigma Supelco Supplier No. 442719
Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 124-19-6
Vapor Pressure ˜0.26 mmHg (25°C)
Density 0.827 g/mL (at 25°C)
Flash Point 64°C
Physical Form Neat
Linear Formula CH3(CH2)7 CHO
MDL Number MFCD00007030
Boiling Point 93°C (23 mmHg)
Synonym Aldehyde C9; Nonyl aldehyde; Pelargonaldehyde
Type Food and Cosmetic Component Standard
Quantity 1000 mg
Formula Weight 142.24
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.